Skip to Content
Merck
All Photos(1)

Key Documents

G6048

Sigma-Aldrich

Galactokinase human

recombinant, expressed in E. coli

Sign Into View Organizational & Contract Pricing

Select a Size

200 μG
€587.00

€587.00


Estimated to ship on08 April 2025


Request a Bulk Order

Select a Size

Change View
200 μG
€587.00

About This Item

Enzyme Commission number:
UNSPSC Code:
12352204
NACRES:
NA.54

€587.00


Estimated to ship on08 April 2025


Request a Bulk Order

recombinant

expressed in E. coli

Quality Level

form

solution

specific activity

≥1400 unit/μg protein

mol wt

42 kDa

shipped in

dry ice

storage temp.

−70°C

Biochem/physiol Actions

Galactokinase catalyzes the phosphorylation of αD-galactose to produce galactose-1-phosphate as part of the Leloir pathway[1].

Physical properties

N-terminal GST-tagged 42 kDa full length protein

Unit Definition

One unit will convert 1.0 picomole of galactose to galactose-1-phosphate per minute at pH 7.4 at 30 °C.

Physical form

Supplied as a solution in 40 mM Tris-HCl, pH 8.0, 110 mM NaCl, 2.2 mM KCl, 20% glycerol, 3 mM DTT and 10-250 mM imidazole.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dariusz Abramczyk et al.
Eukaryotic cell, 11(3), 334-342 (2012-01-03)
The regulation of the Saccharomyces cerevisiae GAL genes in response to galactose as a source of carbon has served as a paradigm for eukaryotic transcriptional control over the last 50 years. Three proteins--a transcriptional activator (Gal4p), an inhibitor (Gal80p), and
Shivani Malik et al.
Nucleic acids research, 40(8), 3348-3363 (2011-12-27)
Recently, we have demonstrated a predominant association of Rad26p with the coding sequences but not promoters of several GAL genes following transcriptional induction. Here, we show that the occupancy of histone H2A-H2B dimer at the coding sequences of these genes
Aurélie Egert et al.
Plant & cell physiology, 53(5), 921-929 (2012-03-23)
Galactokinase (GALK, EC 2.7.1.6) is a cytosolic enzyme with a wide occurrence across the taxonomic kingdoms. It catalyzes the phosphorylation of α-d-galactose (Gal) to α-d-Gal-1-P. The cytotoxicity of free (unphosphorylated) Gal is well documented in plants and causes marked defects.
Erin M Green et al.
Molecular biology of the cell, 23(7), 1367-1375 (2012-02-11)
The genome is nonrandomly organized within the nucleus, but it remains unclear how gene position affects gene expression. Silenced genes have frequently been found associated with the nuclear periphery, and the environment at the periphery is believed to be refractory
Nils Janzen et al.
Archives of medical research, 42(7), 608-612 (2011-12-14)
Galactokinase (GALK) deficiency is an autosomal recessive disorder causing cataract formation that can be prevented or mitigated by early diagnosis and galactose-restricted diet. The aim of this retrospective study was to explore whether GALK-deficiency meets the criteria for neonatal mass

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service