Skip to Content
Merck
All Photos(1)

Key Documents

M8146

Sigma-Aldrich

Methyl-β-D-thiogalactoside

Synonym(s):

Methyl-1-thio-β-D-galactopyranoside, TMG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14O5S
CAS Number:
Molecular Weight:
210.25
Beilstein:
81583
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

synthetic

Assay

≥98% (TLC)

form

powder

technique(s)

thin layer chromatography (TLC): suitable

color

white

solubility

water: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

CS[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C7H14O5S/c1-13-7-6(11)5(10)4(9)3(2-8)12-7/h3-11H,2H2,1H3/t3-,4+,5+,6-,7+/m1/s1

InChI key

LZFNFLTVAMOOPJ-PZRMXXKTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The uptake of methyl-β-D-thiogalactoside (TMG), lactose, and glucose is maintained by the phosphoenolpyruvate-dependent phosphotransferase system.

Application

Methyl-β-D-thiogalactoside has been used in a study to analyze inducers of the E.coli lac repressor system. [1] It has also been used in a study that investigated the utilization of lactose by Streptococcus faecalis. [2]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D L Wyborski et al.
Nucleic acids research, 19(17), 4647-4653 (1991-09-11)
Although the inducible prokaryotic lac repressor system has been successfully adapted for control of gene expression in mammalian cells, little information is available on the pharmacokinetics of beta-galactoside inducers in mammalian cells for optimizing this system. These studies directly measure
Christopher T Oberg et al.
Journal of medicinal chemistry, 51(7), 2297-2301 (2008-03-06)
Anionic O2 derivatives of methyl 3-deoxy-3-(4-methylbenzamido)-1-thio-beta-D-galactopyranoside have been synthesized as inhibitors against galectin-3. The sulfate, H-phosphonate, and benzyl phosphate derivatives showed an increased affinity as compared to the parent unsubstituted galactopyranoside. Modeling revealed arginine-144 being pinched by the C3 benzamide
β-D-phosphogalactoside galactohydrolase of Streptococcus faecalis and the inhibition of its synthesis by glucose
Heller, K. and R. Roschenthaler
Canadian Journal of Microbiology, 24, 512-519 (1979)
Anja Marbach et al.
Journal of biotechnology, 157(1), 82-88 (2011-11-15)
Most commonly used expression systems in bacteria are based on the Escherichia coli lac promoter. Furthermore, lac operon elements are used today in systems and synthetic biology. In the majority of the cases the gratuitous inducers IPTG or TMG are
E H Heuberger et al.
European journal of biochemistry, 267(1), 228-234 (1999-12-22)
A carbohydrate-transport assay was developed that does not require isotopically labelled substrates, but allows transport reactions to be followed spectrophotometrically. It makes use of a membrane system (hybrid membranes or proteoliposomes) bearing the transport system of interest, and a pyrroloquinoline

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service