Monoclonal Anti-Gravin (mouse IgG2a isotype) is derived from the hybridoma JP74 produced by the fusion of mouse myeloma cells (X63Ag8.653) and splenocytes from BALB/c mice immunized with fractionated membrane cortex proteins from human neurons [cell lines NT2-N and SK-N-BE(2)]. Gravin protein is a myristoylated high molecular weight cytoplasmic protein expressed in endothelial cells, cultured fibroblasts and osteosarcoma cells and highly expressed in the brain.
Specificity
Monoclonal Anti-Gravin antibody is specific for human, rat and mouse gravin (approx. 300 kDa).
Immunogen
fractionated membrane cortex proteins from human neuron cell lines NT2-N and SK-N-BE(2).
Application
Anti-Gravin antibody has been used in western blotting immunofluorescence.
Monoclonal Anti-Gravin antibody is suitable for use in immunoblotting (using mpkCCD cells ). The product may also be used for immunocytochemistry and immunoprecipitation.
Biochem/physiol Actions
Gravin (or AKAP12) is a scaffolding kinase that functions as a tumor suppressor. This kinase interacts with PKC, PKA and other cell cycle signaling proteins to modulate cell division, growth, differentiation and inflammatory responses. The expression of gravin isoforms (α, β, and γ) is regulated by tissue-specific promoters. Gravin also acts as an autoantigen in myasthenia gravis patients .
Physical form
Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Frontiers in bioscience : a journal and virtual library, 7, d1782-d1797 (2002-07-23)
Scaffolding proteins are thought to facilitate the efficiency and specificity of enzyme/substrate reactions by coordinating their interaction along a cytoskeletal infrastructure in a spacial and temporal manner. Rodent SSeCKS, and its human orthologue, Gravin, seem to function as tumor suppressors
The Journal of biological chemistry, 279(53), 56014-56023 (2004-10-22)
A Kinase Anchoring Protein 12 (AKAP12; also known as src-suppressed C kinase substrate (SSeCKS) and Gravin) is a multivalent anchoring protein with tumor suppressor activity. Although expression of AKAP12 has been examined in a number of contexts, its expression control
Previous studies in yeast have supported the view that post-transcriptional regulation of protein abundances may be more important than previously believed. Here we ask the question: "In a physiological regulatory process (the response of mammalian kidney cells to the hormone
Journal of gastroenterology, 44(9), 897-911 (2009-05-27)
As the premalignant lesion of human esophageal adenocarcinoma (EAC), Barrett's esophagus (BE) is characterized by intestinal metaplasia in the normal esophagus (NE). Gene expression profiling with microarray and serial analysis of gene expression (SAGE) may help us understand the potential
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.