Skip to Content
Merck
All Photos(1)

Key Documents

F8890

Sigma-Aldrich

Folic acid

BioReagent, suitable for plant cell culture, ≥97%

Synonym(s):

PteGlu, Pteroyl-L-glutamic acid, Vitamin M

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H19N7O6
CAS Number:
Molecular Weight:
441.40
Beilstein:
100781
EC Number:
MDL number:
UNSPSC Code:
12352200
eCl@ss:
34058014
PubChem Substance ID:

product line

BioReagent

Assay

≥97%

technique(s)

cell culture | plant: suitable

application(s)

agriculture

SMILES string

NC(N1)=NC(C2=C1N=CC(CNC3=CC=C(C(N[C@@H](CCC(O)=O)C(O)=O)=O)C=C3)=N2)=O

InChI

1S/C19H19N7O6/c20-19-25-15-14(17(30)26-19)23-11(8-22-15)7-21-10-3-1-9(2-4-10)16(29)24-12(18(31)32)5-6-13(27)28/h1-4,8,12,21H,5-7H2,(H,24,29)(H,27,28)(H,31,32)(H3,20,22,25,26,30)/t12-/m0/s1

InChI key

OVBPIULPVIDEAO-LBPRGKRZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Folic acid is a vitamin that is reduced to dihydrofolate in vivo. Dihydrofolate is further reduced tetrahydrofolate, a methyl group shuttle required for the de novo synthesis of purines, thymidylic acid, and certain amino acids.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tor A Strand et al.
Pediatrics, 135(4), e918-e926 (2015-03-25)
Folate and vitamin B-12 are important for growth. Many children in low- and middle-income countries have inadequate intakes of these nutrients. We undertook a randomized, placebo controlled double-blind trial in 1000 North Indian children, 6 to 35 months of age
Tihomir Tomasić et al.
Bioorganic & medicinal chemistry letters, 19(1), 153-157 (2008-11-19)
Mur ligases catalyze the biosynthesis of the UDP-MurNAc-pentapeptide precursor of peptidoglycan, an essential polymer of bacterial cell-wall. They constitute attractive targets for the development of novel antibacterial agents. Here we report on the synthesis of a series of 2,4-diaminoquinazolines, quinazoline-2,4(1H,3H)-diones
R Wendel Naumann et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 31(35), 4400-4406 (2013-10-16)
Vintafolide (EC145) is a folic acid-desacetylvinblastine conjugate that binds to the folate receptor (FR), which is expressed on the majority of epithelial ovarian cancers. This randomized phase II trial evaluated vintafolide combined with pegylated liposomal doxorubicin (PLD) compared with PLD
Roy Lardenoije et al.
Progress in neurobiology, 131, 21-64 (2015-06-15)
Epigenetics is a quickly growing field encompassing mechanisms regulating gene expression that do not involve changes in the genotype. Epigenetics is of increasing relevance to neuroscience, with epigenetic mechanisms being implicated in brain development and neuronal differentiation, as well as
Breige McNulty et al.
The American journal of clinical nutrition, 98(1), 92-98 (2013-05-31)
Supplementation with folic acid (FA) is recommended worldwide before and during early pregnancy because of its proven effect in preventing neural tube defects, but the role of FA after the 12th gestational week (GW) is much less clear. We investigated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service