![Folic acid European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/115/827/a5b3d8bc-3227-4e52-bf51-9ccd37463b7c/640/a5b3d8bc-3227-4e52-bf51-9ccd37463b7c.png)
Folic acid
![Folic acid European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/115/827/a5b3d8bc-3227-4e52-bf51-9ccd37463b7c/640/a5b3d8bc-3227-4e52-bf51-9ccd37463b7c.png)
Synonym(s):
PteGlu, Pteroyl-L-glutamic acid, Vitamin M, Folic acid
Empirical Formula (Hill Notation):
C19H19N7O6
CAS Number:
Molecular Weight:
441.40
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
API Family
Reference Material Type
Special Grade
Technique
Showing 1-8 of 8
Compare
Product Number
Description
Pricing