Skip to Content
Merck
All Photos(1)

Key Documents

F4051

Sigma-Aldrich

Proteasome Fraction II from rabbit

5 mg/mL, Reticulocyte cell extract, solution

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€654.00

€654.00


Estimated to ship on10 April 2025



Select a Size

Change View
5 MG
€654.00

About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€654.00


Estimated to ship on10 April 2025


Quality Level

form

solution

concentration

5 mg/mL

storage temp.

−70°C

General description

Manufactured for Sigma by Boston Biochem., Inc.

Application

Proteasome Fraction II is ideal for demonstrating ubiquitin- and ATP-dependent degradation/conjugation of radiolabeled or immunodetectable substrates. Isopeptidase inhibitor and a proteasome inhibitor are highly recommended for the accumulation of ubiquitin protein conjugates. Typical assay concentration for FII is 0.5 - 4 mg/ml.

Biochem/physiol Actions

Essentially ubiquitin and ATP free. Contains E1, most E2s, UCHs, and the proteasome 20S and 26S isoforms.
Protein fraction of a cell extract that binds to anion-exchange resin.

Physical form

Solution in 25 mM HEPES, pH 7.6.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Hershko et al.
The Journal of biological chemistry, 261(26), 11992-11999 (1986-09-15)
In order to gain insight into the mechanisms that determine the selectivity of the ubiquitin proteolytic pathway, the protein substrate binding site of the ubiquitin-protein ligase system was identified and examined. Previous studies had shown that the ligase system consists
A Hershko et al.
The Journal of biological chemistry, 258(13), 8206-8214 (1983-07-10)
By affinity chromatography of a crude reticulocyte extract on ubiquitin-Sepharose, three enzymes required for the conjugation of ubiquitin with proteins have been isolated. One is the ubiquitin-activating enzyme (E1), which is covalently linked to the affinity column in the presence

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service