Skip to Content
Merck
All Photos(1)

Key Documents

EP5895150004

Rotor for Eppendorf® 5920R Centrifuges

FA-6x50, aerosol tight, to hold, 6 x 5/50 mL tubes, lid

Synonym(s):

Lid for Eppendorf® FA-45-48-11 Rotor

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€2,550.00

€2,550.00


Check Cart for Availability


Select a Size

Change View
1 EA
€2,550.00

About This Item

UNSPSC Code:
41103909
NACRES:
NB.82

€2,550.00


Check Cart for Availability

feature

lid
to hold 6 x 5/50 mL tubes
aerosol tight
fixed angle rotor

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 5895150004 (Global)

parameter

18,500 × g max. RCF

technique(s)

centrifugation: suitable

compatibility

for use with centrifuge models 5920R

Looking for similar products? Visit Product Comparison Guide

General description

  • Eppendorf® 5920R Centrifuge rotors are high-speed rotors for applications in 5 mL to 50 mL tubes.
  • Aerosol-tight for safe centrifugation of potentially hazardous materials.
  • Includes QuickLock rotor lid for ergonomic fast lid locking.
  • 45° borehole angle minimizes pellets smear.
  • Compact, low weight design for easy handling.
  • Made of chemically resistant anodized aluminum.
  • Rotor, lid and adapters are autoclavable.
The large Centrifuge 5920 R is a benchtop centrifuge that combines exceptional capacity with a compact and ergonomic design. It has similar dimensions to refrigerated competitor models in the 3-liter segment, but has an outstanding capacity of up to 4 x 1,000 mL or 52 x 50 mL tubes. Optimize high-throughput centrifugation and make your daily work easier.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service