Skip to Content
Merck
All Photos(1)

Key Documents

EP5810725003

Eppendorf® Swing-bucket rotor A-4-81-MTP

incl. 4 MTP buckets

Synonym(s):

Lid for Eppendorf® FA-45-48-11 Rotor

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€3,100.00

€3,100.00


Check Cart for Availability


Select a Size

Change View
1 EA
€3,100.00

About This Item

UNSPSC Code:
41103909
NACRES:
NB.82

€3,100.00


Check Cart for Availability

feature

to hold 4 x MTP/flex buckets
incl. 4 MTP buckets

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 5810725003

parameter

4,000 max. RCF (3,220 xg)

technique(s)

centrifugation: suitable

compatibility

for use with centrifuge models 5804/5804R/5810/5810R

General description

  • A versatile swing-bucket rotor (A-4-81-MTP) accommodates 2 plate buckets and 2 rectangular tube buckets facilitating mixed loading.
  • Rotor, lid and adapters are autoclavable at 121 °C, 20 min.
  • Optional aerosol-tight caps are available for the safe centrifugation of hazardous samples.
  • Rotor lifetime: max. 100,000 cycles.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service