Key Documents
EP5702737003
Eppendorf® Adapter for 85/100 mL Round Bucket
holds Vacutainer and Falcon tubes 2-7 ml, pack of 2 ea
Select a Size
2 EA
€197.00
€197.00
Check Cart for Availability
Select a Size
Change View
2 EA
€197.00
About This Item
UNSPSC Code:
41103911
NACRES:
NB.82
€197.00
Check Cart for Availability
feature
holds Vacutainer and Falcon tubes 2-7 ml
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5702737003
technique(s)
centrifugation: suitable
compatibility
for use with 100 mL round buckets
General description
adapter for use with Eppendorf 85/100 mL round bucket
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)