Skip to Content
Merck
All Photos(2)

Key Documents

EP022363263

Eppendorf® Safe-Lock micro test tubes

capacity 1.5 mL, green, pkg of 500 ea

Synonym(s):

eppendorfs

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121703
Pricing and availability is not currently available.

material

conical bottom
green
polypropylene (push fit
)

sterility

non-sterile

feature

RCF 30,000  × g

packaging

pkg of 500 ea

manufacturer/tradename

Eppendorf® 022363263

capacity

1.5 mL

diam.

10.7 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Eppendorf Safe-Lock Tubes, 1.5 mL, Eppendorf Quality, green, 500 tubes
  • Hinged Eppendorf Safe-Lock lid prevents unintentional lid opening during incubation and storage
  • g-Safe®: Depending on Safe-Lock Tube variant (see manual) exceptional centrifugation stability up to 30,000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples
  • Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results
  • Polypropylene provides high resistance to chemicals, mechanical stress and temperature extremes
  • Precise lid sealing for minimal evaporation rates during long-term storage
  • Large frosted lid and surface on the side for easy labeling
  • Ensured functionality from -86 °C to 100 °C
  • Autoclavable when open (121 °C, 20 min)
  • Also available in purity grades of lot-certified PCR clean, Biopur® and Eppendorf Forensic DNA Grade
  • Also available in variants Eppendorf Protein LoBind or Eppendorf DNA LoBind for maximum recovery of valuable samples and in amber for light-sensitive samples

Features and Benefits

Eppendorf Safe-Lock Tubes offer special protection against unintentional opening and completely eminate any danger of leaks during incubation and storage

Legal Information

Biopur is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
g-Safe is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service