Skip to Content
Merck
All Photos(1)

Key Documents

EP0030123611

Eppendorf® Safe-Lock microcentrifuge tubes

Forensic DNA Grade, volume 1.5 mL, pack of 500 tubes (10 bags x 50 pcs.)

Synonym(s):

eppendorf tubes, microcentrifuge tubes, microfuge tubes

Sign Into View Organizational & Contract Pricing

Select a Size

500 EA
€32.50

€32.50


Check Cart for Availability


Select a Size

Change View
500 EA
€32.50

About This Item

UNSPSC Code:
41121703
NACRES:
NB.22

€32.50


Check Cart for Availability

material

cap (Safe-Lock lid)
colorless
polypropylene
polypropylene

sterility

non-sterile

feature

Forensic DNA Grade

packaging

pack of 500 tubes (10 bags x 50 pcs.)

manufacturer/tradename

Eppendorf® 0030123611

parameter

30,000 × g max. RCF

volume

1.5 mL

foreign activity

PCR inhibitors, human DNA, DNase, RNase

PCR inhibitors, human DNA, DNase, RNase, free

Looking for similar products? Visit Product Comparison Guide

General description

Eppendorf® Forensic DNA Grade products comply with the sophisticated requirements of forensic DNA laboratories such as promoting automation to minimize the contact between the handler and product, adhering to strict cleaning procedures and protocols, and positive air pressure that blocks the intrusion of dust. It mainly focuses on the prevention of contamination in forensic DNA analysis. Eppendorf Forensic DNA Grade products are suitable for sample processing, DNA extraction, sample storage, and polymerase chain reaction (PCR) setup.

Features and Benefits

  • Unintentional lid opening during incubation and storage is prevented due to the hinged
  • Eppendorf Safe-Lock lid Tubes are manufactured without biocides, plasticizers, or slip agents that studies have shown can leach out and contaminate samples
  • The tubes are composed of polypropylene that shows resistance to chemicals, extreme temperatures, and mechanical stress
  • The lid sealing enables minimal evaporation during long-term storage of samples
  • Autoclavable at at 121°C for 20 min
  • Functions between -86 °C to 100 °C
  • Large frosted lid and surface on the side for labeling
  • Centrifugation stability up to 30,000 × g averts sample loss due to tube breakage and provides safety while using hazardous samples
Tubes snap closed and stay closed. Flat top with writing surface for positive sample identification

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service