Key Documents
EP0030132890
Eppendorf® Masterclear cap strips and real-time PCR tube strips
10 x 12 strips
Select a Size
120 EA
€297.00
€297.00
Check Cart for Availability
Select a Size
Change View
120 EA
€297.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.11
€297.00
Check Cart for Availability
sterility
non-sterile
manufacturer/tradename
Eppendorf® 0030132890
technique(s)
PCR: suitable
working volume
0.1 mL
General description
Eppendorf® Masterclear Cap Strips and real-time PCR Tube Strips:
Real-time PCR Tube Strips:
  • White wells for better reflection.
  • High mechanical stability.
  • Extremely thin walls for optimal heat transfer.
Masterclear Cap Strips:
  • Inverted dome to avoid scratching of the optic surface.
  • Inverted dome to reduce volume of the tubes.
  • Optimized for maximum light transmission.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)