Skip to Content
Merck
All Photos(1)

Key Documents

BR781327

BRAND® PCR tubes and caps, strips of 8

domed caps

Synonym(s):

BIO-CERT, Cap Strips, PCR, PCR Tube Strips, PCR Tubes, Tube Strips, Tubes

Sign Into View Organizational & Contract Pricing

Select a Size

250 EA
€231.00

€231.00


Check Cart for Availability


Select a Size

Change View
250 EA
€231.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.22

€231.00


Check Cart for Availability

material

colorless tube
domed cap
polypropylene tube

sterility

non-sterile

feature

neck thread push fit
domed caps

packaging

pack of 250 ea
pkg of (250 tube strips + 250 cap strips)

manufacturer/tradename

BRAND 781327

technique(s)

PCR: suitable

capacity

0.2 mL

working volume

0.2 mL

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

PCR 8- and 12-tube strips offer a flexible solution for your PCR or qPCR. Domed caps provide extra sealing pressure via the lid of the thermal cycler. Flat caps are required for qPCR. Attached cap strips can be easily opened and closed with one hand. Separate cap and film strips are suitable for automation, as they won′t obstruct robotics. No matter which cap you choose, all variants seal the vessels reliably, effectively protecting your samples from evaporation and contamination. The extra-thin walls allow for even heat transfer during the process of heating and cooling for short cycle time.

Application

PCR, qPCR (flat caps only)

Features and Benefits

  • High flexibility with a wide range of cap types and film strips
  • Reduced evaporation losses thanks to tightly closing cap strips
  • Rapid heat transfer through thin vessel walls
  • Numbered wells for clear identification
  • Free of: DNA, RNA, DNase, RNase, pyrogens, PCR inhibitors

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service