Key Documents
EP0030106318
Eppendorf® UVette® routine pack, reclosable box
pkg of 200 ea
Synonym(s):
Staining, Staining Accessories
Select a Size
200 EA
€146.00
€146.00
Check Cart for Availability
Select a Size
Change View
200 EA
€146.00
About This Item
UNSPSC Code:
41123304
NACRES:
SE.13
€146.00
Check Cart for Availability
packaging
pkg of 200 ea
manufacturer/tradename
Eppendorf® 0030106318
Features and Benefits
  • UVette® in routine packs are ideal for routine applications.
  • These packs allow a safe storage of the UVettes in a reclosable box, while offering convenient access to each cuvette for easy filling.
  • UV transparent plastic, free of fluoropolymers and other halogenated hydrocarbons.
  • Two built-in optical path lengths in a single cuvette - turn the UVette by 90° to change from 10 mm and 2 mm.
  • Volume markings for 500 μL and 1,000 μL.
  • Deep-lying optical window prevents scratches.
  • Tapered cuvette base for optimal filling, frosted grip for labeling.
  • Ideal for use in the Eppendorf BioPhotometer and Eppendorf BioSpectrometer, adapters available for use with other common spectrophotometers.
  • Routine pack for convenient access to each UVette and safe storage in a reclosable box.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
UVette is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)