Key Documents
EP0030106300
Eppendorf® UVette®, disposable single seated cUVette®s
pkg of 80 ea
Synonym(s):
Eppendorf UVette
Select a Size
80 EA
€69.60
€69.60
Check Cart for Availability
Select a Size
Change View
80 EA
€69.60
About This Item
UNSPSC Code:
41121813
NACRES:
NB.44
€69.60
Check Cart for Availability
packaging
pkg of 80 ea
manufacturer/tradename
Eppendorf® 0030106300
Features and Benefits
  • Eppendorf® UVette® disposable single seated cuvettes are perfect for applications outside of the UV range, such as colorimetric protein assays (Bradford, Lowry, etc.), determining of the optical density of bacterial cultures (OD600 methods), and kinetic and fluorescence measurements.
  • Individually packaged.
  • PCR-clean and protein-free quality.
  • UV transparent plastic, free of fluoropolymers and other halogenated hydrocarbons.
  • Two built-in optical path lengths in a single cuvette - just turn the UVette 90° to change from 10 mm and 2 mm.
  • Volume markings for 500 μL and 1000 μL.
  • Individually packaged and certified PCR clean and protein-free quality for sensitive (e.g. RNA) and valuable samples.
  • Deep-lying optical window prevents scratches.
  • Tapered cuvette base for optimal filling, frosted grip for labeling.
  • Ideal for use in the Eppendorf BioPhotometer and Eppendorf BioSpectrometer, adapters available for use with other common spectrophotometers.
  • Routine pack for convenient access to each UVette and safe storage in a reclosable box.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
UVette is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)