Skip to Content
Merck
All Photos(1)

Key Documents

EP0030078705

Eppendorf® ep Dualfilter T.I.P.S.® SealMax

PCR clean, sterile, tip volume × L 0.5-20 μL × 46 mm, light gray, case of 960 tips (10 racks x 96 tips)

Synonym(s):

pipetting tips

Sign Into View Organizational & Contract Pricing

Select a Size

960 PKG
€202.00

€202.00


Check Cart for Availability


Select a Size

Change View
960 PKG
€202.00

About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
volume:
0.5—20 μL
sterility:
sterile (PCR clean/sterile/pyrogen-free)
sterile
feature:
PCR clean
Low binding, filter, RNA or DNA free, pcr, low retention etc.
packaging:
case of 960 tips (10 racks x 96 tips)
rack of 960 ea (10 racks of 96 tips)

€202.00


Check Cart for Availability

sterility

sterile (PCR clean/sterile/pyrogen-free)
sterile

feature

PCR clean
Low binding, filter, RNA or DNA free, pcr, low retention etc.

packaging

case of 960 tips (10 racks x 96 tips)
rack of 960 ea (10 racks of 96 tips)

tip volume × L

0.5-20 μL × 46 mm

volume

0.5—20 μL

color

light gray

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The new violet and white filter for the ep Dualfilter T.I.P.S.® SealMax filter tips combine a reliable contamination protection for your pipette from your sample liquid with virtually 100% protection from aerosols1 and biomolecules.

This new Dualfilter closes immediately when it comes into contact with the overpipetted sample without inhibiting valuable material. Additionally drops and splashes are retained by the white layer, the violet layer forms the highly-effective barrier against sample liquid.

Further information and certification on specific sample recovery features, PCR inhibition and aerosol protection are available on your local Eppendorf website.

Product Applications:
  • DNA applications (e.g. PCR)
  • RNA applications (e.g. Gene expression analysis)
  • Protein applications (e.g. Antibody Research)
  • Cell Culture applications (e.g. Media)
  • Applications with radio actives
  • All applications which use aerosol1-binding liquids

Product Features:
  • Reliable protection from liquids
  • Reliable protection from aerosols1 and biomolecules
  • No PCR inhibiting additives
  • Protection of your pipette from aggressive liquids when over-aspiration occurs
  • Available in PCR clean / Sterile (sterile and pyrogen-free)
  • Continuous inspection of each production lot by an independent laboratory guarantees high purity and quality

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
ep Dualfilter T.I.P.S. is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service