Skip to Content
Merck
All Photos(1)

Documents

EHU130111

Sigma-Aldrich

MISSION® esiRNA

targeting human CTCF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AACAGCAGGAGGGTCTGCTATCAGAGGTTAATGCAGAGAAAGTGGTTGGTAATATGAAGCCTCCAAAGCCAACAAAAATTAAAAAGAAAGGTGTAAAGAAGACATTCCAGTGTGAGCTTTGCAGTTACACGTGTCCACGGCGTTCAAATTTGGATCGTCACATGAAAAGCCACACTGATGAGAGACCACACAAGTGCCATCTCTGTGGCAGGGCATTCAGAACAGTCACCCTCCTGAGGAATCACCTTAACACACACACAGGTACTCGTCCTCACAAGTGCCCAGACTGCGACATGGCCTTTGTGACCAGTGGAGAATTGGTTCGGCATCGTCGTTACAAACACACCCACGAGAAGCCATTCAAGTGTTCCATGTGCGATTACGCCAGTGTAGAAGTCAGCAAATTAAAACGTCACATTCGCTCTCATACTGGAGAGCGTCCGTTTCAGTGCAGTTTGTGCAGTTATGCCAGCAGGGACACATACAAGCTGAAAAGGCACATGA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nathan A Damaschke et al.
Clinical epigenetics, 12(1), 80-80 (2020-06-07)
The chromatin insulator CCCTC-binding factor (CTCF) displays tissue-specific DNA binding sites that regulate transcription and chromatin organization. Despite evidence linking CTCF to the protection of epigenetic states through barrier insulation, the impact of CTCF loss on genome-wide DNA methylation sites
Zhengfei Shan et al.
Journal of cellular and molecular medicine, 23(5), 3130-3139 (2019-03-16)
The present research focuses on the influence of CCCTC-binding factor (CTCF) on prostate cancer (PC) via the regulation of the FoxO signalling pathway. A bioinformatics analysis was conducted to screen out target genes for CTCF in LNCaP cells and to
Lena N Lupey-Green et al.
Virology, 507, 220-230 (2017-04-30)
The Epstein Barr virus (EBV) genome persists in infected host cells as a chromatinized episome and is subject to chromatin-mediated regulation. Binding of the host insulator protein CTCF to the EBV genome has an established role in maintaining viral latency
Li Sun et al.
Theranostics, 10(26), 11950-11962 (2020-11-19)
Rationale: Mesenchymal stem cells (MSCs) have been the focus of many studies because of their abilities to modulate immune responses, angiogenesis, and promote tumor growth and metastasis. Our previous work showed that gastric cancer MSCs (GCMSCs) promoted immune escape by
Francisco Puerta Martínez et al.
Journal of virology, 88(13), 7389-7401 (2014-04-18)
Human cytomegalovirus (HCMV) gene expression during infection is highly regulated, with sequential expression of immediate-early (IE), early (E), and late (L) gene transcripts. To explore the potential role of chromatin regulatory factors that may regulate HCMV gene expression and DNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service