Skip to Content
Merck
All Photos(1)

Key Documents

EHU109561

Sigma-Aldrich

MISSION® esiRNA

targeting human PSMD4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GAAGGTGGCAAGATGGTGTTGGAAAGCACTATGGTGTGTGTGGACAACAGTGAGTATATGCGGAATGGAGACTTCTTACCCACCAGGCTGCAGGCCCAGCAGGATGCTGTCAACATAGTTTGTCATTCAAAGACCCGCAGCAACCCTGAGAACAACGTGGGCCTTATCACACTGGCTAATGACTGTGAAGTGCTGACCACACTCACCCCAGACACTGGCCGTATCCTGTCCAAGCTACATACTGTCCAACCCAAGGGCAAGATCACCTTCTGCACGGGCATCCGCGTGGCCCATCTGGCTCTGAAGCACCGACAAGGCAAGAATCACAAGATGCGCATCATTGCCTTTGTGGGAAGCCCAGTGGAGGACAATGAGAAGGATCTGGTGAAACTGGCTAAACGCCTCAAGA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pere Dosta et al.
Cardiovascular engineering and technology, 12(1), 114-125 (2021-01-22)
Endothelial cell (EC) dysfunction underlies the pathology of multiple disease conditions including cardiovascular and pulmonary diseases. Dysfunctional ECs have a distinctive gene expression profile compared to healthy ECs. RNAi therapy is a powerful therapeutic approach that can be used to
Lei Xu et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(10), 13194-13210 (2020-12-16)
Ablation of miR-144/451 disrupts homeostasis of erythropoiesis. Myc, a protooncogenic protein, is essential for erythroblast proliferation but commits rapid downregulation during erythroid maturation. How erythroblasts orchestrate maturation processes through coding and non-coding genes is largely unknown. In this study, we
Ai-Guo Ma et al.
The Kaohsiung journal of medical sciences, 35(10), 591-597 (2019-06-05)
Proteasome 26S subunit non-ATPase 4 (PSMD4) is an important proteasome ubiquitin receptor and plays a key role in endoplasmic reticulum stress (ERS). However, the study of PSMD4 in esophageal cancer (EC) is relatively rare. Here, we found that the expression
Padma Murthi et al.
Cells, 9(4) (2020-04-16)
We reported earlier that an anti-inflammatory small peptide receptor-formyl peptide receptor-2 (FPR2) was significantly decreased in placentas from third trimester pregnancies complicated with fetal growth restriction (FGR), compared to placentas from uncomplicated control pregnancies, suggesting FPR2 may play a role
Emma-Kate Loveday et al.
The Journal of general virology, 96(Pt 1), 30-39 (2014-09-23)
A common critical cellular event that many human enveloped viruses share is the requirement for proteolytic cleavage of the viral glycoprotein by furin in the host secretory pathway. For example, the furin-dependent proteolytic activation of highly pathogenic (HP) influenza A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service