Skip to Content
Merck
All Photos(5)

Key Documents

HPA035503

Sigma-Aldrich

Anti-LRRC15 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-LIB, Anti-Leucine rich repeat containing 15

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Check Cart for Availability


Select a Size

Change View
100 μL
€505.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€505.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:200

immunogen sequence

NWLLLNQPRLGTDTVPVCFSPANVRGQSLIIINVNVAVPSVHVPEVPSYPETPWYPDTPSYPDTTSVSSTTELTSPVEDYTDLTTIQVTDDRSVW

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LRRC15(131578)

General description

Leucine-rich repeat-containing protein 15 (LRRC15), a type I transmembrane protein, is majorly expressed in stromal fibroblasts. It is a member of the leucine-rich repeat superfamily and is devoid of intracellular signaling domains. The LRRC15 gene is mapped to human chromosome 3q29. The levels of LRRC15 are higher in mineralized tissues.

Immunogen

leucine rich repeat containing 15 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Leucine-rich repeat-containing protein 15 (LRRC15) participates in the cell-cell and cell-extracellular matrix interactions. It favors osteogenic differentiation under in vitro and in vivo conditions. LRRC15 shows elevated expression in various solid tumors and also in high-grade, aggressive breast and prostate tumors.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78604

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jim O'Prey et al.
Journal of virology, 82(12), 5933-5939 (2008-04-04)
Adenoviruses for gene or oncolytic therapy are under development. Notable among these strategies is adenoviral delivery of the tumor suppressor p53. Since all therapeutics have limitations in certain settings, we have undertaken retroviral suppressor screens to identify genes conferring resistance
Paul A Reynolds et al.
Genes & development, 17(17), 2094-2107 (2003-08-19)
Desmoplastic small round cell tumor (DSRCT) is defined by a chimeric transcription factor, resulting from fusion of the N-terminal domain of the Ewing's sarcoma gene EWS to the three C-terminal zinc fingers of the Wilms' tumor suppressor WT1. Although DNA-binding
Yuejun Wang et al.
Stem cell research & therapy, 9(1), 65-65 (2018-03-11)
Mesenchymal stem cells (MSCs) are a reliable resource for bone regeneration and tissue engineering, but the molecular mechanisms of differentiation remain unclear. The tumor antigen 15-leucine-rich repeat containing membrane protein (LRRC15) is a transmembrane protein demonstrated to play important roles
James W Purcell et al.
Cancer research, 78(14), 4059-4072 (2018-05-17)
Progress in understanding tumor stromal biology has been constrained in part because cancer-associated fibroblasts (CAF) are a heterogeneous population with limited cell-type-specific protein markers. Using RNA expression profiling, we identified the membrane protein leucine-rich repeat containing 15 (LRRC15) as highly

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service