Skip to Content
Merck
All Photos(1)

Key Documents

EHU070981

Sigma-Aldrich

MISSION® esiRNA

targeting human E2F1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€197.00
50 μG
€349.00

€197.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
20 μG
€197.00
50 μG
€349.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€197.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ACCCCTCTCCTCTGTCTCCAGAAGCTTCTAGCTCTGGGGTCTGGCTACCGCTAGGAGGCTGAGCAAGCCAGGAAGGGAAGGAGTCTGTGTGGTGTGTATGTGCATGCAGCCTACACCCACACGTGTGTACCGGGGGTGAATGTGTGTGAGCATGTGTGTGTGCATGTACCGGGGAATGAAGGTGAACATACACCTCTGTGTGTGCACTGCAGACACGCCCCAGTGTGTCCACATGTGTGTGCATGAGTCCATGTGTGCGCGTGGGGGGGCTCTAACTGCACTTTCGGCCCTTTTGCTCTGGGGGTCCCACAAGGCCCAGGGCAGTGCCTGCTCCCAGAATCTGGTGCTCTGACCAGGCCAGGTGGGGAGGCTTTGGCTGGCTGGGCGTGTAGGACGGTGAGAGCACTTCT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qian Li et al.
Cell death & disease, 11(9), 757-757 (2020-09-17)
Despite the ubiquitous mechanical cues at both spatial and temporal dimensions, cell identities and functions are largely immune to the everchanging mechanical stimuli. To understand the molecular basis of this epigenetic stability, we interrogated compressive force-elicited transcriptomic changes in mesenchymal
Juan Liu et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 37(2), 2673-2682 (2015-09-26)
Cancerous inhibitor of protein phosphatase 2A (CIP2A) is a recently identified oncoprotein. Here, we investigated its role in the formation of multidrug resistance (MDR) of cervical adenocarcinoma in vitro and in vivo. MTT assay showed that knockdown of CIP2A expression
Jung-Jyh Hung et al.
Experimental hematology & oncology, 1(1), 18-18 (2012-12-06)
The transcription factor E2F1 has been implicated in cell cycle control and DNA damage response. Paradoxically, E2F1 can promote apoptosis and function as tumor suppressor. In non-small cell lung cancer (NSCLC), there are conflicting data for clinical significance of E2F1
Wen Luo et al.
Scientific reports, 6, 27904-27904 (2016-06-11)
miR-17 family microRNAs (miRNAs) are crucial for embryo development, however, their role in muscle development is still unclear. miR-20a-5p and miR-20b-5p belong to the miR-17 family and are transcribed from the miR-17~92 and miR-106a~363 clusters respectively. In this study, we
Yuan He et al.
Life sciences, 252, 117656-117656 (2020-04-15)
Diabetes is considered as one of the important risks in the progression of Hepatocellular carcinoma(HCC). Ribosome binding protein 1 (RRBP1), a rough endoplasmic reticulum protein, plays an essential role in diabetes and various cancer. E2F transcription factor 1 (E2F1), an

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service