Skip to Content
Merck
All Photos(1)

Key Documents

EHU016891

Sigma-Aldrich

MISSION® esiRNA

targeting human SERPINE1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€197.00
50 μG
€349.00

€197.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
20 μG
€197.00
50 μG
€349.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€197.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTGGTTCTGCCCAAGTTCTCCCTGGAGACTGAAGTCGACCTCAGGAAGCCCCTAGAGAACCTGGGAATGACCGACATGTTCAGACAGTTTCAGGCTGACTTCACGAGTCTTTCAGACCAAGAGCCTCTCCACGTCGCGCAGGCGCTGCAGAAAGTGAAGATCGAGGTGAACGAGAGTGGCACGGTGGCCTCCTCATCCACAGCTGTCATAGTCTCAGCCCGCATGGCCCCCGAGGAGATCATCATGGACAGACCCTTCCTCTTTGTGGTCCGGCACAACCCCACAGGAACAGTCCTTTTCATGGGCCAAGTGATGGAACCCTGACCCTGGGGAAAGACGCCTTCATCTGGGACAAAACTGGAGATGCATCGGGAAAGAAGAAACTCCGAAGAAAAGAATTTTAGTGTTAATGACTCTTTCTGAAGGAAGAGAAGACATTTGCCTTTTGTTAAAAGATGGTAAACCAGATCTGTCTCCAAGACCTTGGCCTCTCCTTGGAGGACCTTT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyeon-Joon Kong et al.
International journal of oncology, 58(1), 111-121 (2020-12-29)
Serpin family E member 1 (SERPINE1), a serine proteinase inhibitor, serves as an important regulator of extracellular matrix remodeling. Emerging evidence suggests that SERPINE1 has diverse roles in cancer and is associated with poor prognosis. However, the mechanism via which SERPINE1 is induced
Amanda M Rampersaud et al.
Molecular human reproduction, 26(11), 850-865 (2020-09-09)
A critical component of early human placental development includes migration of extravillous trophoblasts (EVTs) into the decidua. EVTs migrate toward and displace vascular smooth muscle cells (SMCs) surrounding several uterine structures, including spiral arteries. Shallow trophoblast invasion features in several
En-Dong Zhu et al.
PloS one, 9(8), e106049-e106049 (2014-08-30)
Gastric cancer is one of the most common malignant diseases worldwide. Emerging evidence has shown that microRNAs (miRNAs) are associated with tumor development and progression. Our previous studies have revealed that H. pylori infection was able to induce the altered
Mike R Wilson et al.
Cell reports, 33(6), 108366-108366 (2020-11-12)
Endometriosis affects 1 in 10 women and is characterized by the presence of abnormal endometrium at ectopic sites. ARID1A mutations are observed in deeply invasive forms of the disease, often correlating with malignancy. To identify epigenetic dependencies driving invasion, we
Ruozhi Zhao et al.
Journal of leukocyte biology, 95(6), 941-949 (2014-02-06)
Diabetes mellitus accelerates the development of atherosclerotic cardiovascular diseases. Monocyte adhesion is an early cellular event of atherogenesis. Elevated levels of glyLDL were common in diabetic patients. Our previous studies indicated that HSF1 and p22-phox (a subunit of the NOX

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service