Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 50 μL | Check Cart for Availability | € 531,00 |
About This Item
€ 531,00
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
species reactivity
human
technique(s)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 3-16 μg/mL using human heart tissue
UniProt accession no.
1 of 1
This Item | |||
|---|---|---|---|
| conjugate unconjugated | conjugate - | conjugate unconjugated | conjugate unconjugated |
| Quality Level 200 | Quality Level 200 | Quality Level 100 | Quality Level 100 |
| biological source rabbit | biological source rabbit | biological source rabbit | biological source mouse |
| antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form purified immunoglobulin |
| species reactivity human | species reactivity human | species reactivity human, pig, chicken, bovine | species reactivity human |
| storage temp. −70°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
Immunogen
Application
Immunohistochemistry (1 paper)
Biochem/physiol Actions
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Instructions
Global Trade Item Number
| SKU | GTIN |
|---|---|
| E9780-50UL | 04061837832246 |
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service


