Skip to Content
Merck
All Photos(3)

Key Documents

E3651

Sigma-Aldrich

Anti-Endothelin Receptor A antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-ET-A, Anti-ET1-Specific Endothelin Receptor

Sign Into View Organizational & Contract Pricing

Select a Size

0.05 ML
€390.00
0.2 ML
€852.00

€390.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
0.05 ML
€390.00
0.2 ML
€852.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€390.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human, pig, chicken, bovine

technique(s)

immunohistochemistry: 1:100 using rat brain sections or rat lung paraffin embedded sections
western blot: 1:200 using rat brain membranes and ECL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EDNRA(1909)
rat ... Ednra(24326)

Specificity

The epitope is highly conserved in human, swine, bovine and chicken ET-A.

Immunogen

synthetic peptide corresponding to amino acids 413-426 of rat ET-A (with additional N-terminal cysteine).

Application

Anti-Endothelin Receptor A antibody produced in rabbit is suitable for immunohistochemistry at a dilution of 1:100 using rat brain sections or rat lung paraffin embedded sections and western blotting at a dilution of 1:200 using rat brain membranes and ECL.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

Endothelin receptor type A (ETA, ETAR) is a human G protein-coupled receptor that mediates vasoconstriction of blood vessels. It is involved in tumor cell invasion, survival and metastasis. ETAR mediates the promotion of ET-1 on colon cancer cell survival, invasion and MMP-2 expression by a PI3K-mediated mechanism. It plays a critical role in colon cancer metastasis.

Physical form

Lyophilized from a solution of phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shaolin Nie et al.
Molecular carcinogenesis, 53 Suppl 1, E85-E91 (2013-07-03)
The endothelin (ET)-1/endothelin A receptor (ETAR) axis is reportedly involved in tumor cell invasion, survival, and metastasis. However, the role of ETAR in colon cancer metastasis and the underlying mechanisms have not been defined. In the present study, we assessed
S Hori et al.
Endocrinology, 130(4), 1885-1895 (1992-04-01)
Endothelins (ETs) are very potent vasoconstrictive peptides and have diverse functions in both vascular and nonvascular tissues. This investigation concerns the tissue distribution and cellular localization of rat mRNAs encoding two different subtypes of ET receptors (ETA and ETB). We
Shaoqing He et al.
Investigative ophthalmology & visual science, 56(10), 6144-6161 (2015-09-24)
A growing body of evidence suggests that the vasoactive peptides endothelins (ETs) and their receptors (primarily the ETB receptor) are contributors to neurodegeneration in glaucoma. However, actions of ETs in retinal ganglion cells (RGCs) are not fully understood. The purpose
H Y Lin et al.
Proceedings of the National Academy of Sciences of the United States of America, 88(8), 3185-3189 (1991-04-15)
By screening a cDNA library derived from the A10 rat vascular smooth muscle cell line for functional expression in COS cells, we have isolated a high-affinity receptor for endothelin 1 (Kd = 476 pM) and endothelin 2. The affinity of
Kyung Jin Jung et al.
Journal of Korean medical science, 26(1), 92-99 (2011-01-11)
Endothelin (ET)-1 and its receptors (ETA and ETB receptor) are present in the central nervous system. ET exerts biological effects on gliogenesis and glial cell functions. In order to define a possible mechanism of ETA receptor signaling, the distribution of

Questions

  1. Could you provide the molecular weight (MW) of the antibody E3651 Anti-Endothelin Receptor A?

    1 answer
    1. The synthetic peptide corresponds to amino acid residues 413-426 of rat ET-A (Accession P26684) and has multiple isoforms. There are multiple glycosylation and phosphorylation sites (Accession P26684). After performing a western blot, you will observe multiple bands, with sizes approximately around 48 KDa, 28 KDa, and so on. For further details on the protein size and its different isoforms, please visit the UniProt website and refer to the following link: UniProt - Accession P26684.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service