Skip to Content
Merck

Skip To

A8426

Albumin, glycated bovine

lyophilized powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
50 mg
Check Cart for Availability
€ 2.160,00
€ 1.836,00

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.26
MDL number:
eCl@ss:
32160409
Biological source:
bovine
Assay:
≥90% protein basis (biuret)
Form:
lyophilized powder
Technique(s):
cell culture | mammalian: suitable
Concentration:
≥90% protein (biuret)

€ 1.836,00

List Price€ 2.160,00Save 15%
Web-Only Promotion

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine

Quality Segment

assay

≥90% protein basis (biuret)

form

lyophilized powder

composition

Protein, ~95% biuret

origin

USA origin

concentration

≥90% protein (biuret)

extent of labeling

1-5 mol hexose (as fructosamine) per mol albumin

technique(s)

cell culture | mammalian: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

bovine ... ALB(280717)

Physical form

Lyophilized powder containing citrate buffer salts

Preparation Note

Glycated in vitro

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
N8159A3264A9418
technique(s)

cell culture | mammalian: suitable

technique(s)

immunoblotting: suitable

technique(s)

hybridization: suitable, microbiological culture: suitable, protein quantification: suitable

technique(s)

cell culture | mammalian: suitable

origin

USA origin

origin

-

origin

-

origin

USA origin

biological source

bovine

biological source

bovine

biological source

sheep

biological source

bovine

assay

≥90% protein basis (biuret)

assay

-

assay

≥97% (agarose gel electrophoresis)

assay

≥96% (agarose gel electrophoresis)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters