Skip to Content
Merck
All Photos(3)

Key Documents

DCAS9P300

Sigma-Aldrich

Sigma CRISPR dCas9-p300 Activator Expression Plasmid

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€250.00

€250.00


Usually ships in 4 to 6 weeks.

Request a Bulk Order

Select a Size

Change View
1 EA
€250.00

About This Item

UNSPSC Code:
41106610
NACRES:
NA.51

€250.00


Usually ships in 4 to 6 weeks.

Request a Bulk Order

recombinant

expressed in E. coli

Quality Level

packaging

vial of 50 μL

concentration

20 ng/μL in TE buffer; DNA (1μg of plasmid DNA)

application(s)

CRISPR

shipped in

dry ice

storage temp.

−20°C

General description

A gene activation system based on a fusion of inactive Cas9 or dCas9 to the catalytic histone acetyltransferase (HAT) core domain of the human E1A-associated protein p300. The dCas9-p300 CRISPR Gene Activator with the p300 domain represents a distinct mechanism of action relative to dCas9-VP64 or other similar gene activation motifs. The p300 histone acetyltransferase protein opens a transcriptional highway by releasing the DNA from its heterochromatin state and allowing for continued and robust gene expression by the endogenous cellular machinery. The Cas9 expression plasmids use the CMV promoter for strong transient expression of Cas9. Alternate promoters can be substituted by replacement of CMV using MluI and NheI. Also, the Cas9 expression plasmids can be linearized using XbaI for T7-based mRNA production.

Application

  • Functional Genomics/Target ValidationEpigenetic Modification
  • Transcriptional Activation

Features and Benefits

  • The Sigma CRISPR dCas9p300 plasmid co-expresses p300-HAT and GFP, to easily monitor delivery and expression in your target cell type (figure 1)gRNAs can successfully direct nuclease-deficient Cas9 (dCas9) fused to p300 HAT catalytic domain to increase levels of histone acetylation and endogenous gene expressionThe dCas9-p300 histone acetylation approach represents a distinct mechanism of action relative to dCas9-VP64 or other similar gene activation motifs

Components

1 vial containing 1ug of dCas9-p300 plasmid.

Linkage

To order U6-gRNA plasmids, please visit sigma.com/crisprs


Click here for more information on dcas9-p300 Gene Activator Expression Plasmid.

Other Notes

Must be used with U6-gRNA only expressing plasmid.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Sigma has developed a gene activation system based on a fusion of dCas9 to the catalytic histone acetyltransferase (HAT) core domain of the human E1A-associated protein p300.

Related Content

Sigma-Aldrich® Advanced Genomics is the leading provider of gene editing and silencing technologies including CRISPR, Cas9, synthetic guide RNA (sgRNA), and Zinc Finger Nuclease (ZFN).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service