Skip to Content
Merck
All Photos(2)

Key Documents

D4567

Sigma-Aldrich

Anti-DNMT1 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-DNA methyltransferase

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
€477.00

€477.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 μL
€477.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€477.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~180 kDa

species reactivity

rat, mouse, human

technique(s)

indirect immunofluorescence: 1:100-1:200 using mouse COS-7 cells
microarray: suitable
western blot: 1:1,000-1:2,000 using nuclear extracts of human HEK 293T cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DNMT1(1786)
mouse ... Dnmt1(13433)

General description

Human DNA methyltransferase 1 (Dnmt1) is a 1616 amino acids protein; the N-terminal two-thirds of the protein is considered to be the regulatory domain, while the C-terminal region contains the catalytic domain.
In humans, the gene encodes a DNA (cytosine-5)-methyltransferase with 1616 amino acids. The N-terminal contains the regulatory domain, while the C-terminal region contains the catalytic domain. Two isoforms of DNMT1 have been isolated, DNMT1a and DNMT1b, the difference residing in 16 extra amino acids within the latter.

Immunogen

synthetic peptide corresponding to amino acids 1581-1595 of rat DNMT1, conjugated to KLH via an N-terminal added lysine residue. The sequence is conserved in human and mouse.

Application

Anti-DNMT1 antibody produced in rabbit has been used in western blot analysis, and staining for confocal laser scanning microscopy.

Biochem/physiol Actions

DNA methyltransferase 1 (Dnmt1) is part of different complexes; it can establish a repressive transcription complex consisting of the histone deacetylase 4 (HDAC4), DNA methyltransferase 1-associated Protein (DMAP1) and DNMT1 itself. Consistent with the role of DNA methylation in gene silencing, DNMT1 can associate with HDAC1 and MECP2 (methyl CpG binding protein 2).
DNMT1 protein is important for the maintenance of methylation, as well as for the novo methylation activities occurring in somatic cells of vertebrates. It establishes a repressive transcription complex at replication foci with histone deacetylase HDAAC4 and DMAP1.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Depolarization induces downregulation of DNMT1 and DNMT3 in primary cortical cultures
Sharma RP, et al.
Epigenetics, 3(2), 74- 80 (2008)
Laccaic Acid A Is a Direct, DNA-competitive Inhibitor of DNA Methyltransferase 1
Fagan RL, et al.
The Journal of Biological Chemistry, 288(33), 23858-23858 (2013)
Rajiv P Sharma et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 35(10), 2009-2020 (2010-07-16)
The study of CpG methylation of genomic DNA in neurons has emerged from the shadow of cancer biology into a fundamental investigation of neuronal physiology. This advance began with the discovery that catalytic and receptor proteins related to the insertion
Rajiv P Sharma et al.
Epigenetics, 3(2), 74-80 (2008-06-10)
DNA methylation in post-mitotic neurons is reported to serve a variety of functions from survival during development to the consolidation of memory. Of particular interest with regards neuronal functioning is the change in site-specific methylation of a variety of gene
I Rhee et al.
Nature, 404(6781), 1003-1007 (2000-05-09)
Hypermethylation is associated with the silencing of tumour susceptibility genes in several forms of cancer; however, the mechanisms responsible for this aberrant methylation are poorly understood. The prototypic DNA methyltransferase, DNMT1, has been widely assumed to be responsible for most

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service