Skip to Content
Merck
All Photos(18)

Documents

ZRB5535

Sigma-Aldrich

Anti-SOX-9 Antibody, clone 2B10, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Transcription factor SOX-9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2B10, monoclonal
recombinant monoclonal

description

2B10 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 56.14 kDa
observed mol wt ~65 kDa

species reactivity

mouse, human

species reactivity (predicted by homology)

rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable using 1:100
immunohistochemistry: suitable using 1:100
western blot: suitable using 1:1,000

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... SOX9(6662)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 2B10 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects human and murine SOX-9. It targets an epitope with in 24 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 24 amino acids from the C-terminal region of human SOX-9.

Application

Anti-SOX-9, clone 2B10, ZooMAb is a recombinant Rabbit Monoclonal antibody that detects Sox-9 and has been tested for use in Immunocytochemistry, Immunohistochemistry, and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected SOX-9 in HepG2 cell lysate.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected SOX-9 in SW480 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected SOX-9 in E16.5 mouse embryo, mouse nasal cavity, and developing mouse gut tissue sections.

Target description

Transcription factor SOX-9 (UniProt P48436; also known SOX-9) is encoded by the SOX9 gene (Gene ID: 6662) in human. SOX family of genes are related to the mammalian sex determining gene SRY. These genes contain sequences encoding for the HMG-box domains that mediate sequence-specific DNA binding. SOX genes encode transcriptional regulators involved in cell fate decision during development and the control of diverse developmental processes. The highly complex group of SOX genes cluster at about 40 genomic loci and at least 30 SOX genes have been identified. SOX-9 contains one HMG box DNA-binding domain and acts as a key regulator of developmental processes, including male sex determination, normal skeletal development, chondrogenesis, neurogenesis, and neural crest development. SOX-9 is acts downstream of sonic hedgehog signaling and is shown to be essential for the maintenance of multipotent neural stem cells in embryonic and adult central nervous system (CNS). It is not detected in neuroblasts of the adult subependymal zone (SEZ); however, it is expressed by transit amplifying cells and ependymal cells in the same region. SOX-9 is also shown to be essential for the ′switch′ from neurogenesis to gliogenesis in caudal regions of the developing CNS. Up-regulation of SOX-9 is reported in cancers of various origins, including breast, colon, prostate, and lung, where its expression correlates with malignancy and patient survival. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Scott, CE et al. (2010). Nat. Neurosci. 13(10); 1181-89).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

30 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Follow this short antibody reconstitution protocol and see how easy it is to reconstitute ZooMAb® recombinant antibodies.

Related Content

Greener ZooMAb® antibodies offer sustainable options with low-waste packaging and BSA-free formulation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service