Skip to Content
Merck
All Photos(1)

Key Documents

D2693

Sigma-Aldrich

Anti-phospho-DARPP32 (pThr75) antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution, sufficient for 10 blots

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
€619.00

€619.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 VIAL
€619.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€619.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

usage

sufficient for 10 blots

mol wt

antigen 32 kDa

species reactivity

mouse, human

technique(s)

dot blot: 1:1,000
western blot: 1:1,000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PPP1R1B(84152)
mouse ... Ppp1r1b(19049)

General description

DARPP32 is a phosphoprotein with a molecular mass of 32kDa. It is regulated by dopamine (DA) and cAMP and is associated with dopaminoceptive neurons bearing D-1 receptors in the basal ganglia. DARPP32 is a target for the actions of dopamine.

Immunogen

synthetic phosphopeptide corresponding to amino acids residues surrounding the DARPP32 phosphorylated on threonine 75.

Application

Anti-phospho-DARPP32 (pThr75) antibody produced in rabbit is suitable for dot blot and western blot analysis at a working dilution of 1:1000 using rat caudate lysates.

Biochem/physiol Actions

Dopamine D1 receptor stimulation enhances cAMP formation, resulting in the phosphorylation of DARPP32. Phosphorylation of DARPP32 plays an important role in the regulation of dopaminergic neurotransmission. The activity of this protein has an important role in the actions of alcohol, caffeine and Prozac®.

Target description

DARPP32 phosphorylated on Thr75 acts as an inhibitor of PKA.

Physical form

Solution at 100 μL in 10 mM HEPES, pH 7.5, 150 mM NaCl, 100 μg/mL BSA, and 50% glycerol.

Other Notes

The labeling by the antibody to DARPP32 Thr75 is markedly increased by okadaic acid treatment.

Legal Information

Prozac is a registered trademark of Eli Lilly and Co.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J A Bibb et al.
Nature, 402(6762), 669-671 (1999-12-22)
The physiological state of the cell is controlled by signal transduction mechanisms which regulate the balance between protein kinase and protein phosphatase activities. Here we report that a single protein can, depending on which particular amino-acid residue is phosphorylated, function
Per Svenningsson et al.
Science (New York, N.Y.), 302(5649), 1412-1415 (2003-11-25)
Three distinct classes of drugs: dopaminergic agonists (such as D-amphetamine), serotonergic agonists (such as LSD), and glutamatergic antagonists (such as PCP) all induce psychotomimetic states in experimental animals that closely resemble schizophrenia symptoms in humans. Here we implicate a common
R E Maldve et al.
Nature neuroscience, 5(7), 641-648 (2002-06-18)
The medium spiny neurons of the nucleus accumbens receive both an excitatory glutamatergic input from forebrain and a dopaminergic input from the ventral tegmental area. This integration point may constitute a locus whereby the N-methyl-D-aspartate (NMDA)-subtype of glutamate receptors promotes
Maria Lindskog et al.
Nature, 418(6899), 774-778 (2002-08-16)
Caffeine has been imbibed since ancient times in tea and coffee, and more recently in colas. Caffeine owes its psychostimulant action to a blockade of adenosine A(2A) receptors, but little is known about its intracellular mechanism of action. Here we
A A Fienberg et al.
Science (New York, N.Y.), 281(5378), 838-842 (1998-08-07)
Dopaminergic neurons exert a major modulatory effect on the forebrain. Dopamine and adenosine 3',5'-monophosphate-regulated phosphoprotein (32 kilodaltons) (DARPP-32), which is enriched in all neurons that receive a dopaminergic input, is converted in response to dopamine into a potent protein phosphatase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service