Skip to Content
Merck
All Photos(1)

Documents

D041

Sigma-Aldrich

S(+)-Propylnorapomorphine hydrochloride

solid

Synonym(s):

S(+)-NPA hydrochloride, S(+)10,11-Dihydroxy-N-n-propylnoraporphine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21NO2 · HCl
CAS Number:
Molecular Weight:
331.84
MDL number:
UNSPSC Code:
12352210
PubChem Substance ID:
NACRES:
NA.77

form

solid

Quality Level

color

off-white

solubility

H2O: moderately soluble
ethanol: soluble (Solutions should be freshly prepared.)

storage temp.

2-8°C

SMILES string

Cl.[H][C@@]12Cc3ccc(O)c(O)c3-c4cccc(CCN1CCC)c24

InChI

1S/C19H21NO2.ClH/c1-2-9-20-10-8-12-4-3-5-14-17(12)15(20)11-13-6-7-16(21)19(22)18(13)14;/h3-7,15,21-22H,2,8-11H2,1H3;1H/t15-;/m0./s1

InChI key

PCOQOGIDTIFQAM-RSAXXLAASA-N

General description

Dissolve in oxygen-free boiled water containing 0.1% sodium metabisulfite or other antioxidant. Solutions should be freshly prepared.

Biochem/physiol Actions

Limbic-selective dopamine antagonist.

Caution

Store tightly sealed at 4 °C; subject to rapid oxidation. Packed under argon; reseal bottle under argon or nitrogen to maximize stability.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J L Neumeyer et al.
Journal of medicinal chemistry, 26(4), 516-521 (1983-04-01)
The enantiomers [(S)-(+) and (R)-(-)] of N-n-propylnorapomorphine (NPA) were synthesized. (R)-NPA was obtained by the acid-catalyzed rearrangement of N-n-propylnormorphine. (R)-NPA also was converted to (RS)-N-n-propylnorapomorphine dimethyl ether by dehydrogenation of the 10,11-O,O'-dimethyl ether of (R)-NPA with 10% palladium on carbon
R F Cox et al.
The Journal of pharmacology and experimental therapeutics, 247(1), 355-362 (1988-10-01)
Prompted by conflicting reports of both agonist and antagonist properties of the S-(+)-enantiomer of the potent dopamine agonist R-(-)-N-n-propylnorapomorphine (NPA), we carried out extracellular, single unit recording studies to compare the effects of both enantiomers on substantia nigra and ventral

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service