CLS7249
Corning® TC treated 1536 well microplates
white, high base, w/o lid, sterile
All Photos(1)
About This Item
Recommended Products
material
polystyrene plate
square wells
white
sterility
sterile
feature
lid: no
packaging
case of 50 ea
manufacturer/tradename
Corning 7249
size
1536 wells
well volume
12.5 μL
well working volume
1-8 μL
color
white
Related Categories
Features and Benefits
- Superior performance compared to other available commercial microplates: lower CV values, excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
- Optimized for automation with bar coding and standard readers
- Surface treated for optimal cell attachment
- Flat bottom wells with lid are compatible with automated dispensers including the PerkinElmer® FlexDrop
- Sterile and certified nonpyrogenic
Legal Information
Corning is a registered trademark of Corning, Inc.
PerkinElmer is a registered trademark of PerkinElmer Inc.