CLS430641
Corning® cell culture flasks
surface area 75 cm2, canted neck, cap (vented)
Synonym(s):
culture flask, flask cell culture, tissue culture flask
All Photos(1)
About This Item
Recommended Products
material
canted neck
cap (vented)
polystyrene
rectangular flask
sterility
sterile
feature
graduations
packaging
case of 100
manufacturer/tradename
Corning 430641
maximum volume
60 mL
surface area
75 cm2
working volume
15-22.5 mL
General description
Available in a variety of sizes, designs and cap styles to meet your needs.
- Manufactured from optically clear virgin polystyrene
- Treated for optimal cell attachment
- Printed with lot numbers for ease in traceability
- 100% integrity tested
- Sterilized by gamma irradiation and certified nonpyrogenic
Legal Information
Corning is a registered trademark of Corning, Inc.
recommended
Product No.
Description
Pricing
replaced by