Skip to Content
Merck
All Photos(1)

Key Documents

CLS3898

Corning® 96 Well Clear Polystyrene Microplate

V-bottom clear, polystyrene, bag of 25, non-sterile, lid: no

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
€413.00

€413.00


Check Cart for Availability


Select a Size

Change View
100 EA
€413.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.24
material:
V-bottom clear
clear bottom
polystyrene
size:
96 wells

€413.00


Check Cart for Availability

material

V-bottom clear
clear bottom
polystyrene

sterility

non-sterile

feature

lid: no
skirt

packaging

bag of 25

manufacturer/tradename

Corning 3898

size

96 wells

well working volume

200 μL

color

clear

suitability

suitable for (cell cuture)

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Corning® 96 Well Clear Polystyrene Microplate without lids are made of virgin polystyrene.
  • Recommended working volume of up to 75 to 200 μL.
  • Processed to improve hydrophilicity for hemagglutination and similar assays.
  • Conforms to standard microplate footprint and dimensions.
  • Corona discharge treated hydrophilic surface and negatively charged.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abdisalan M Noor et al.
BMC infectious diseases, 11, 121-121 (2011-05-17)
Countries aiming for malaria elimination require a detailed understanding of the current intensity of malaria transmission within their national borders. National household sample surveys are now being used to define infection prevalence but these are less efficient in areas of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service