CLS353112
Corning® Falcon® Cell Culture Flask
capacity 350 mL, straight neck, graduated, 50 ‑ 600 mL, cap, blue vented
Synonym(s):
Corning container
About This Item
Recommended Products
material
polystyrene
feature
cap blue vented
graduated 50 ‑ 600 mL
manufacturer/tradename
Corning 353112
technique(s)
cell culture | mammalian: suitable
capacity
350 mL
Looking for similar products? Visit Product Comparison Guide
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service