Skip to Content
Merck
All Photos(2)

Key Documents

CLS352099

Corning® Falcon® Centrifuge Tubes

capacity 15 mL, cap, dome seal screw, racked, sterile

Synonym(s):

culture tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121703
NACRES:
NB.13
Pricing and availability is not currently available.

material

conical polystyrene

sterility

sterile

feature

cap dome seal screw

manufacturer/tradename

Corning 352099

capacity

15 mL

Looking for similar products? Visit Product Comparison Guide

Application

Liquid Handling

Legal Information

Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service