Skip to Content
Merck
All Photos(1)

Key Documents

CLS3384

Corning® HTS Transwell® 96 well permeable supports

HTS Transwell-96 Permeable Support with 8.0 μm pore polyester membrane, TC-treated, sterile, 8/cs

Synonym(s):

corning transwell, transwell corning

Sign Into View Organizational & Contract Pricing

Select a Size

8 EA
€845.00

€845.00


Estimated to ship on29 March 2025



Select a Size

Change View
8 EA
€845.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
flat bottom wells clear
polyethylene membrane
polystyrene
round wells
insert working volume:
0.08 mL
membrane diam.:
4.26 mm
membrane thickness:
10.0 μm
size:
96 wells
surface area:
0.14 cm2 , cell growth area
well working volume:
0.235 mL

€845.00


Estimated to ship on29 March 2025


material

flat bottom wells clear
polyethylene membrane
polystyrene
round wells

sterility

sterile

feature

lid
skirt
plate format: 96 well

packaging

pack of 4
case of 8

manufacturer/tradename

Corning 3384

insert working volume

0.08 mL

membrane diam.

4.26 mm

membrane thickness

10.0 μm

size

96 wells

surface area

0.14 cm2 , cell growth area

well working volume

0.235 mL

color

clear plate

pore size

8.0 μm pore size

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • HTS Transwell®-96 permeable supports have an array of 96 wells with permeable inserts connected by a rigid robotics-friendly tray
  • Product includes HTS Transwell®-96 permeable support insert, receiver plate (96 wells for growth or assay steps) and 1 sterile lid; 8 sets per case
  • 8.0 μm pores size PET (polyester) membrane
  • Membrane area for cell growth or cell migration is 0.143 cm² which is 20-50% greater than other brands
  • Large apical and basolateral access ports
  • Automation optimized design with multi-channel feeder ports, improved gripping surface and bar coding
  • All membranes are treated for cell attachment
  • Receiver plates are untreated
  • Lid – minimizes evaporation and protects against contamination

Legal Information

Corning is a registered trademark of Corning, Inc.
Transwell is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service