CLS3374
Corning® HTS Transwell® 96 well permeable supports
HTS Transwell-96 Permeable Support with 8.0 μm pore polyester membrane, TC-treated, sterile, 2/cs
Synonym(s):
corning transwell, transwell corning
Select a Size
€222.00
Select a Size
About This Item
polyethylene membrane
polystyrene
€222.00
Recommended Products
material
flat bottom wells clear
polyethylene membrane
polystyrene
sterility
sterile
feature
lid
skirt
plate format: 96 well
packaging
pack of 1
case of 2
manufacturer/tradename
Corning 3374
insert size
0.08 mL
membrane size
4.26 mm
membrane thickness
10.0 μm
size
96 wells
surface area
0.14 cm2 , cell growth area
well working volume
0.235 mL
color
clear plate
pore size
8.0 μm pore size
suitability
suitable for (cell cuture)
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- HTS Transwell®-96 permeable supports have an array of 96 wells with permeable inserts connected by a rigid robotics-friendly tray
- Product includes HTS Transwell®-96 permeable support insert, receiver plate (96 wells for growth or assay steps) and 1 sterile lid; 2 sets percase
- 8.0 μm pores size PET (polyester) membrane
- Membrane area for cell growth or cell migration is 0.143 cm² which is 20-50% greater than other brands
- Large apical and basolateral access ports
- Automation optimized design with multi-channel feeder ports, improved gripping surface and bar coding
- All membranes are treated for cell attachment
- Receiver plates are untreated
- Lid – minimizes evaporation and protects against contamination
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service