Skip to Content
Merck
All Photos(3)

Key Documents

CLL1167

Sigma-Aldrich

A549 Cells GFP-SMAD4

human male lung (Source Disease: Lung carcinoma)

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
€4,090.00

€4,090.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 VIAL
€4,090.00

About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

€4,090.00


Check Cart for Availability

Request a Bulk Order

Product Name

A549 Cells GFP-SMAD4,

biological source

human male lung (Source Disease: Lung carcinoma)

OMIM accession no.

storage temp.

−196°C

Gene Information

human ... SMAD4(4089)

General description

A549 GFP-SMAD4 are lung carcinoma cells from a human 58 year old caucasian male having two distinct ZFN modifications creating two fluorescent transgenes each expressed from their endogenous gene locus.

This cell line was derived from ATCC Catalog No. CCL-185.

Application

This product is a human A549 cell line in which the genomic SMAD4 gene has been endogenously tagged with a Green Fluorescent Protein (GFP) gene using CompoZr® zinc finger nuclease (ZFN) technology. Integration resulted in endogenous expression of the fusion protein in which GFP is attached to the C-terminus of SMAD4. Fluorescence imaging shows SMAD4-GFP expression throughout the cell. Upon activation with a ligand such as transforming growth factor beta (TGFb), the cell line shows redistribution of SMAD4 from the cytoplasm to the nucleus, making it useful for high content screening to identify compounds that modulate this redistribution. This stable cell line was expanded from a single clone. The target′s gene regulation and corresponding protein function are preserved in contrast to cell lines with overexpression via an exogenous promoter.
To learn more, please visit the Cellular Reporter Cell Line webpage

Features and Benefits

Zinc Finger Nuclease (ZFN)-mediated targeted integration of a fluorescent GFP tag into the last coding exon just preceding the stop codon of the SMAD4 gene on chromosome 18q21.2 to create a cell line exhibiting stable expression of the GFP transgene.

The A549 cells are adherent, with a doubling time of approx. 22 hours.

Quality

Tested for Mycoplasma, sterility, post-freeze viability, short terminal repeat (STR) analysis for cell line identification, cytochrome oxidase I (COI) analysis for cell line species confirmation.

Preparation Note

Media Renewal changes two to three times per week.

Rapidly thaw vial by gentle agitation in 37°C water bath (~2 minutes), keeping vial cap out of the water. Decontaminate with 70% ethanol, add 9 mL culture media and centrifuge 125 x g (5-7 minutes). Resuspend in complete culture media and incubate at 37°C in a 5% CO2 atmosphere.

Subculture Ratio: approx. 1:3-1:8

The base medium for this cell line is F12 Ham Medium, Cat. No. N4888. To make the complete growth medium, add the following components to the base medium: fetal bovine serum, Cat. No. F4135, to a final concentration (v/v) of 10% and L-glutamine, Cat. No. G7513, at a final concentration of 2 mM.

Cell freezing medium-DMSO 1X, Cat. No. C6164.

Legal Information

CompoZr is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

6.2 - Infectious substances

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service