Skip to Content
Merck
All Photos(1)

Key Documents

22Rv1

5092802, human prostate, Epithelial

Synonym(s):

22Rv-1 Cells, CWR-22rv1 Cells, CWR22-R1 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

22Rv1, 05092802

biological source

human prostate

growth mode

Adherent aggregates

karyotype

49,XY,del(1)(p10),+i(1)(q10),der(2)t(2;4)(p13;q31)del(2)(q13q33),der(4)t(2;4)(p13;q31),t(6;14)(q15;q32),+7,+12[5]/50,idem,+3[1]

morphology

Epithelial

products

Not specified

receptors

androgen receptor positive

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Xenograft Prostate

Cell Line Description

22Rv1 is a human prostate carcinoma epithelial cell line derived from a xenograft that was serially propagated in mice after castration-induced regression and relapse of the parental, androgen-dependent CWR22 xenograft. The cell line expresses prostate specific antigen (PSA). Growth is weakly stimulated by dihydroxytestosterone and lysates are immunoreactive with androgen receptor antibody by Western blot analysis. Growth is stimulated by epidermal growth factor (EGF) but is not inhibited by transforming growth factor β-1 (TGF β-1). This cell line is tumorigenic in nude mice.

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 10,11
D13S317: 9,12
D16S539: 12
D5S818: 11,12
D7S820: 10,11
THO1: 6,9.3
TPOX: 8
vWA: 15,21

Culture Medium

RPMI 1640 without phenol red (Sigma R7509) + 2mM Glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Split sub-confluent cultures (70-80%) 1:5 to 1:10 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DNA, RNA, cDNA derived from ECACC mammalian cell lines allow screening for genes or expression patterns to identify lines most suitable for specific research.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service