Recommended Products
product name
TGP 54, 94022430
biological source
mouse pancreas
growth mode
Adherent
karyotype
Not specified
morphology
Not specified
products
Somatostatin
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Related Categories
Cell Line Origin
Mouse pancreatic islet cell tumour
Cell Line Description
TGP 54 has been isolated from an islet tumour arising in the pancreas of a transgenic Tg (E1a-1-SV40E)Bri8 mouse. These mice carry a pseudogene construct composed of a pancreatic-specific elastase-1 promoter linked to the SV40T antigen and thereby represent an animal model for pancreatic carcinogenesis. TGP 54 cells are SV40T antigen positive and secrete low amounts of somatostatin, it is one of a series of cells obtained from pancreatic tumours deposited at ECACC.
Application
Study of exocrine pancreas carcinomas, hormone secretion studies.
Culture Medium
DMEM:Ham′s F12 (1:1) + 2mM Glutamine + 10% Foetal Bovine Serum (FBS) (Heat Inactivated).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:3 i.e. seeding at 3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
ECACC pancreatic cancer cell lines for various applications including drug response studies.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service