Skip to Content
Merck
All Photos(1)

Key Documents

RPMI 1846

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

hamster skin

packaging

tube of 5 μg 93120834-DNA-5UG
pkg of vial of cells 93120834-1VL

growth mode

Adherent

karyotype

2n = 44

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Hamster Syrian melanotic melanoma

Cell Line Description

Derived from a spontaneous malignant melanoma in an aged golden Syrian hamster. The cells have the ability to produce melanin in vitro. The TD50 for new-born hamsters is 68 cells.

Application

Tumourigenicity studies

Culture Medium

McCoy′s 5a + 2mM Glutamine + 10-20% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 i.e. seeding at 1.5x1,000,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service