Skip to Content
Merck
All Photos(1)

Documents

MDCK

85011435, dog kidney, Epithelial-like

Synonym(s):

MDCK1 Cells, MDCKI Cells, Madin Darby Canine Kidney Cells, NBL-2 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32190102

product name

MDCK, 85011435

biological source

dog kidney

growth mode

Adherent

karyotype

2n = 78

morphology

Epithelial-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Canine Cocker Spaniel kidney

Cell Line Description

Derived from the kidney of a normal adult Cocker Spaniel in 1958. Useful in viral studies, supports the growth of vesicular exanthema virus of swine, vesicular stomatitis, vaccina, coxsackie B-5, adenoviruses, reoviruses and infectious canine hepatitis. Also known as MDCK (NBL-2).

Application

Virus studies: SVEV, VSV, Vaccinia, reoviruses and inf. canine hepatitis virus

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C. Cells attach firmly and require at least 2 PBS washes prior to addition of trypsin/EDTA.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service