R4-6A2
91032502, Reactivity: Anti-Interferon-γ murine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
biological source
Mouse x Rat Hybridoma
ImmunoDon
DA rat spleen
Myeloma
P3U.1
Reactivity
Anti-Interferon-γ murine
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Mouse P3U.1 x Rat DA spleen
Cell Line Description
The antibody is directed against murine interferon gamma and neutralises its antiviral activity. It also inhibits lymphokine induced murine macrophage tumoricidal activity.
Immunogen
Murine interferon gamma
Application
The isotype of the monoclonal antibody produced by this cell line is rat IgG1.
Culture Medium
DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)
Subculture Routine
Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service