Skip to Content
Merck
All Photos(1)

Documents

NCTC clone 1469

88111403, mouse liver, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

NCTC clone 1469, 88111403

biological source

mouse liver

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Mouse C3H/An liver

Cell Line Description

Derived from mouse liver cell line NCTC 721 which was obtained from normal liver of a 2 day old C3H/An mouse. The cells have been used in biochemical and nutritional studies. The cells are susceptible to VSV (Indiana strain).

Application

Cell suspension systems. Biochemical, nutritional and virus studies. VSV(Indiana)

Culture Medium

NCTC 135 + 2mM Glutamine + 10% Horse Serum (HS)

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:4 using only mechanical techniques, such as scraper, do not use trypsin, 5-10CO2; 37°C. Change media every 3 days. Freeze in 10% glycerol/90% FBS. Cells are shed into the medium giving it a turbid appearan

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service