Skip to Content
Merck
All Photos(1)

Documents

RCP

31626, rat brain, Endothelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

RCP, 00031626

biological source

rat brain

growth mode

Adherent

karyotype

Not specified

morphology

Endothelial

products

Not specified

receptors

Vasopressin V1a, Angiotensin II, Endothelin 1, Bradykinin, Serotonin

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Rat Choroid Plexus

Cell Line Description

The choroid plexus of adult Wistar Rats were dissected from the fourth ventricle to give a primary culture. This primary culture was then immortalized by transfection with the TSori minus adenovirus (a replication deficient adenovirus containing SV40 large T antigen). Sub-cloning of the immortalised cells was performed by selecting for clones which exhibited calcium ion release in response to vasopressin, angiotensin, bradykinin, serotonin or endothelin stimulation. The sub-clone which exhibited the largest response to all five hormones was selected and sub-cultured on to become RCP.

Application

Toxicology, cellular signalling and pharmacology studies

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 1-3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service