The recommended 1X reaction buffer is 20 mM Tris, pH 7.4, 100 mM NaCl, 5 mM MgCl2, 1 mM DTT. The dilution buffer, which is included in this kit, is recommended for further dilution of reconstituted proteins, if required.
CAS9PL
Cas9 Plus Protein
from Streptococcus pyogenes, recombinant, expressed in E. coli, 3X NLS
Synonym(s):
Cas9, SpCas9, SpyCas9
Select a Size
About This Item
Recommended Products
recombinant
expressed in E. coli
Quality Level
Assay
≥95% (SDS-PAGE)
form
lyophilized powder
packaging
pkg of 1 kit (4 components)
application(s)
CRISPR
shipped in
wet ice
storage temp.
−20°C
General description
Application
Features and Benefits
- Enhanced specificity compared to wild type Cas9
- Increased activity compared to enhanced specificity Cas9
- Ready to inject/transfect
Packaging
pkg of 250 μg (≥ 1500 pmol)
Components
- one vial of Cas9 Plus Recombinant Protein
- one vial of Plus Transfection Buffer
- one vial of Reconstitution Buffer
- one vial of Dilution Buffer
Principle
Reconstitution
Other Notes
Check out our other MISSION® Cas9 Proteins at SigmaAldrich.com/CRISPRproteins
Legal Information
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
Guaranteed PURedit™ CRISPR synthetic gRNAs and Cas9 protein offer industry-leading on-site cutting and specificity
Combine guaranteed sgRNAs with our comprehensive range of CRISPR products and tools, including Cas9 and delivery reagents, for efficient genome modification with higher specificity.
Related Content
Explore technology and reagent portfolios for plant breeding workflows, accelerating the development of new crop varieties.
-
Hi, which reaction buffer can I use for Cas9 Plus-sgRNA complex for in vitro assay? I need to check cleavage of DNA in vitro before injections into animals and I didnt find any protocols or reccomendations in datasheets. Can I use dilution buffer?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service