Skip to Content
Merck
All Photos(1)

Key Documents

C9473

Sigma-Aldrich

Complement Serum Standard human

aqueous solution

Synonym(s):

Human Complement Serum

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€472.00

€472.00


Check Cart for Availability


Select a Size

Change View
1 ML
€472.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.61

€472.00


Check Cart for Availability

biological source

human

Quality Level

form

aqueous solution

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

General description

The complement serum standard human is suitable for the determination of CH50 and AP50 complement activities. This allows for a test of the complement activity on the capacity of the serum to lyse red blood cells in vitro and can be used in procedures for genotyping DNA samples of the complement system.

Application

Complement Serum Standard human has been used as a positive control to study the role of the complement system in immunological demyelination of the spinal cord from mammals.[1]

Biochem/physiol Actions

The complement system is an important constituent of innate immunity.[2] It participates in the first line of defense against microbial infections, altered host cells, and removal of immune complexes and injured cells. The complement system comprises more than thirty components such as the plasma proteins produced by the liver, membrane proteins expressed on the cell surface, cytokines, and hormones.[2][3] The complement proteins come together to opsonize the pathogens and initiate a cascade of inflammatory responses serving the immune cells to fight the infection and maintain homeostasis.[2]

Physical form

Supplied as a 0.22 μm filtered solution containing ~ 18 mM citrate, ~22 mM dextrose, ~3 mM phosphate and ~13 mM calcium

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marina Noris et al.
Seminars in nephrology, 33(6), 479-492 (2013-10-29)
Complement is an important component of the innate immune system that is crucial for defense from microbial infections and for clearance of immune complexes and injured cells. In normal conditions complement is tightly controlled by a number of fluid-phase and
J K Dyer et al.
Spinal cord, 43(7), 417-425 (2005-05-18)
Specificity of serum complement component to elicit immunological demyelination. To assess the role of complement components and pathways in experimental immunological demyelination of the adult rat spinal cord. ICORD, University of British Columbia, Vancouver, Canada. We used 32 adult male
Nicolas S Merle et al.
Frontiers in immunology, 6, 262-262 (2015-06-18)
Complement is a complex innate immune surveillance system, playing a key role in defense against pathogens and in host homeostasis. The complement system is initiated by conformational changes in recognition molecular complexes upon sensing danger signals. The subsequent cascade of
Risk Alleles in CFH and ARMS2 Are Independently Associated with Systemic Complement Activation in Age-related Macular Degeneration.
Smailhodzic D., et al.
Ophthalmology (2011)
Jordan E Inglis et al.
Advances in physiology education, 32(4), 317-321 (2008-12-03)
The immune system is a vital physiological component that affords animals protection from disease and is composed of innate and adaptive mechanisms that rely on cellular and dissolved components. The serum complement system is a series of dissolved proteins that

Questions

  1. This product is from a pool of how many donors?

    1 answer
    1. This human serum used to prepare this product is from a pool of approximately 50 donors.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service