Skip to Content
Merck
All Photos(1)

Key Documents

C2787

Sigma-Aldrich

Complement C7 from human serum

≥85% (SDS-PAGE), ≥150,000 C7H50 units/mg protein

Sign Into View Organizational & Contract Pricing

Select a Size

0.1 MG
€487.00

€487.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
0.1 MG
€487.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.75

€487.00


Check Cart for Availability

Request a Bulk Order

biological source

human serum

Quality Level

Assay

≥85% (SDS-PAGE)

form

solution

specific activity

≥150,000 C7H50 units/mg protein

concentration

1 mg/mL in 10 mM sodium phosphate, 150 mM sodium chloride, pH 7.2

technique(s)

activity assay: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... C7(730)

Biochem/physiol Actions

The complement system is a complex cascade involving proteolytic cleavage of serum glycoproteins often activated by cell receptors. This cascade ultimately results in induction of the inflammatory response, phagocyte chemotaxis and opsonization, and cell lysis.

A complex of complements C5b, C6, C7, and C8 mediates the polymerization of up to eighteen C9 molecules into a tube-like membrane attack complex that is inserted into the plasma membrane of unwanted organisms, such as of Gram-negative bacteria and viral infected cells. This channel through the lipid bilayer results in lysis of the cell.

Human Complement C7 is a β-2-globulin, which is composed of a single polypeptide chain. The initial binding of C7 and C6 to C5b is thought to form a stable complex for subsequent binding of C8 and C9. The resulting activated complex is responsible for membrane attack and lysis.

Quality

Functionally active by a sensitive hemolytic assay.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A F Esser
Toxicology, 87(1-3), 229-247 (1994-02-28)
The membrane attack complex of complement is formed by the molecular fusion of the five terminal complement proteins, C5, C6, C7, C8, and C9. While the assembly process on a target membrane and its modulation by restriction factors present on

Questions

  1. I would like to know if the product is a full sequence protein (92.4 kDa, 843 AA)?

    1 answer
    1. This product is the full-length, unmodified, native protein from human serum. Complement C7 is a naturally glycosylated protein composed of a single polypeptide chain of 92,400 Da.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service