Skip to Content
Merck

Skip To

C6062

Nalgene® sealing cap assembly

diam. 20 mm, pack of 2 ea

Synonym(s):

bottle seal, tube sealing device, Nalgene DS3131-0020, plastic cap

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
2 ea
Check Cart for Availability
€ 67,00
€ 60,30

About This Item

NACRES:
NB.11
UNSPSC Code:
41121703
Material:
Tefzel® plug, Viton® O-ring, polypropylene screw closure
Packaging:
pack of 2 ea

€ 60,30

List Price€ 67,00Save 10%
Web-Only Promotion

Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

Tefzel® plug, Viton® O-ring, polypropylene screw closure

packaging

pack of 2 ea

manufacturer/tradename

Nalgene DS3131-0020

diam.

20 mm

Application

Use with recommended tubes and bottles for leakproof service at maximum speed. Also recommended for spinning hazardous materials.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Tefzel is a registered trademark of E. I. du Pont de Nemours and Company
Viton is a registered trademark of E. I. du Pont de Nemours and Company

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C6187C7312C7687
material

Tefzel® plug, polypropylene screw closure, Viton® O-ring

material

Tefzel® plug, Viton® O-ring, polypropylene screw closure

material

polypropylene screw closure, retained silicone gasket

material

polypropylene screw closure, retained silicone gasket

manufacturer/tradename

Nalgene DS3131-0020

manufacturer/tradename

Nalgene DS3131-0024

manufacturer/tradename

Nalgene DS3132-0020

manufacturer/tradename

Nalgene DS3132-0063

packaging

pack of 2 ea

packaging

pack of 2 ea

packaging

pack of 10 ea

packaging

pack of 2 ea

diam.

20 mm

diam.

24 mm

diam.

20 mm

diam.

63 mm


Still not finding the right product?

Explore all of our products under Nalgene® sealing cap assembly




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library