Skip to Content
Merck

Skip To

C5723

Anti-Cytochrome c antibody produced in sheep

whole antiserum

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
MDL number:
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IF, IP, WB
Citations:
6
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

sheep

Quality Segment

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

mol wt

antigen 14.4 kDa

species reactivity

human, rabbit, canine, rat

technique(s)

immunoprecipitation (IP): suitable, indirect immunofluorescence: suitable, western blot: 1:5,000 using MCF-7, Rat-1, MDCK or Jurkat cell extracts

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CYCS(54205)
rat ... Cycs(25309)

General description

Cytochrome c is an electron transport protein released from mitochondria as an early committed event in apoptosis. Cytochrome c and dATP are cofactors for the mammalian apoptosome, which is composed of Apaf-1, Bcl-2, and procaspase 9. When caspase 9 is activated, activation of other caspases follow including the death protease caspase 3. The release of cytochrome c is inhibited by the presence of Bcl-2 on these organelles preventing the initiation of apoptosis. In cells induced by several apoptotic agents (such as UV irradiation, staurosporine, and overexpression of Bax), caspase inhibitors do not prevent cytochrome c release.

Immunogen

rabbit cytochrome c

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB4502234C8114SAB1410987
conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

whole antiserum

antibody form

affinity isolated antibody

antibody form

IgG fraction of antiserum

antibody form

purified immunoglobulin

biological source

sheep

biological source

rabbit

biological source

rabbit

biological source

rabbit

Quality Level

200

Quality Level

100

Quality Level

-

Quality Level

100

Gene Information

human ... CYCS(54205)
rat ... Cycs(25309)

Gene Information

human ... CYCS(54205)

Gene Information

human ... CTNNA1(1495)
rat ... Ctnna1(307505)

Gene Information

human ... CYCS(54205)

species reactivity

human, rabbit, canine, rat

species reactivity

human, rat, mouse

species reactivity

rat, human, canine

species reactivity

human


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters