C213050
DMT-2′O-TBDMS-rC(ac) Phosphoramidite
configured for (ÄKTA® and OligoPilot®)
Synonym(s):
N-acetyl-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-O-[(1,1-dimethylethyl)dimethylsilyl]-cytidine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite]
Select a Size
About This Item
Recommended Products
biological source
non-animal source (no BSE/TSE risk)
Quality Level
product line
Proligo Reagents
Assay
≥99% (31P-NMR)
≥99.0% (reversed phase HPLC)
form
powder
technique(s)
oligo synthesis: suitable
impurities
≤0.3% water content (Karl Fischer)
≤0.5% P(III) Impurities 100-169ppm (31P-NMR)
≤0.5% single Impurity (reversed phase HPLC)
≤3% residual Solvent content
color
white to off-white
λ
conforms (UV/VIS Identity)
suitability
conforms to structure for H-NMR
conforms to structure for LC-MS
compatibility
configured for (ÄKTA® and OligoPilot®)
nucleoside profile
base: cytidine
base protecting group: acetyl
2' protecting group: TBDMS
5' protecting group: DMT
deprotection: fast/standard
storage temp.
−20°C
SMILES string
COc1ccc(cc1)C(OC[C@H]2O[C@H]([C@H](O[Si](C)(C)C(C)(C)C)[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)N3C=CC(NC(C)=O)=NC3=O)(c4ccccc4)c5ccc(OC)cc5
InChI
1S/C47H64N5O9PSi/c1-32(2)52(33(3)4)62(58-30-16-28-48)60-42-40(59-44(43(42)61-63(11,12)46(6,7)8)51-29-27-41(49-34(5)53)50-45(51)54)31-57-47(35-17-14-13-15-18-35,36-19-23-38(55-9)24-20-36)37-21-25-39(56-10)26-22-37/h13-15,17-27,29,32-33,40,42-44H,16,30-31H2,1-12H3,(H,49,50,53,54)/t40-,42-,43-,44-,62?/m1/s1
InChI key
QKWKXYVKGFKODW-YOEDQOPESA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Features and Benefits
- Industry standard 2′O-TBDMS protective group
- Consistent lot-to-lot purity and performance
- Compatible with deprotection methods based on methylamine or AMA
- Standard RNA phosphoramidites provide excellent coupling results when used with ETT or BTT as an activator; best results are obtained with Activator 42
- Capping with standard acetic anhydride capping reagent rather than with fast Deprotection Cap A
- Manufactured under a certified ISO 9001 quality system
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service